Print…
Colunas Purospher® STAR RP-18 "endcapped" UHPLC
Fornecedor: Merck
1.50650.0001 1.50651.0001 1.50645.0001 1.50646.0001 1.50654.0001 1.50648.0001 1.50653.0001 1.50649.0001 1.50648.1003
1.50650.0001EA 526 EUR
1.50650.0001 1.50651.0001 1.50645.0001 1.50646.0001 1.50654.0001 1.50648.0001 1.50653.0001 1.50649.0001 1.50648.1003
Colunas Purospher® STAR RP-18 "endcapped" UHPLC
Cromatografia, Colunas
Available with optimised 2 μm particulate silica, these UHPLC columns, based on Purospher® STAR RP-18 endcapped, are ideal for ultra-fast applications, where resolution, sensitivity and sample throughput are crucial. They are the first choice for high throughput screening and QC analyses, process monitoring, method development and LC-MS applications. Due to its balanced selectivity, Purospher® STAR RP-18 endcapped UHPLC columns cover almost all demanding separations with tailing-free chromatograms. Purospher® STAR RP-18 endcapped 3 μm columns are recommended for difficult samples whereMais informação do produto