Print…
Reagent bottles, with screw caps
Fornecedor: Merck
1.10392.0001
1.10392.0001EA 163 EUR
1.10392.0001
Reagent bottles, with screw caps
Frascos Frascos de Vidro
Glass, amber.
Amber coloured to protect light-sensitive materials.