Print…
Pronase E (from Streptomyces griseus)
Fornecedor: Merck
1.07433.0001 1.07433.0005
1.07433.0001EA 154 EUR
1.07433.0001 1.07433.0005
Pronase E (from Streptomyces griseus)
Enzimas
Pronase E is a mixture of proteolytic enzymes that is obtained from Streptomyces griseus. It can be used in the in diagnostics in histopathology as it hydrolyses proteins into individual amino acids.