Print…
Sistemas de recolha de solventes
Fornecedor: Merck
1.01114.0001 1.01123.0001 1.06710.0001
1.01114.0001EA 1690 EUR
1.01114.0001 1.01123.0001 1.06710.0001
Sistemas de recolha de solventes
Bombas Bombas de Tambor
Merck offers a broad variety of withdrawal systems that are tailor-made for barrels and drums. They are designed to ensure safe, easy and efficient solvent withdrawal-even of residual quantities-while meeting all relevant safety regulations.
The integrated solvent supply systems directly connect the solvent to the instrument or product line where it is needed. We provide systems for manual pressure build-up as well as inert gas pressurising, supplemented by a comprehensive selection of reducers, adapters and couplings that allow easy interconnection of all components. This way the user canMais informação do produto