Print…

Anti-PD1 Rabbit Monoclonal Antibody [clone: NAT105]

Fornecedor: Abcam
AB216352-20UL AB216352-1ML
ABCAAB216352-20ULEA 275 EUR
ABCAAB216352-20UL ABCAAB216352-1ML
Anti-PD1 Rabbit Monoclonal Antibody [clone: NAT105]
Anticorpos
Rabbit monoclonal [NAT105] to PD1 - Chimeric.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PD1
Clonality: Monoclonal
Clone: NAT105
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR